Merck
  • Analysis of endocrine disrupters and related compounds in sediments and sewage sludge using on-line turbulent flow chromatography-liquid chromatography-tandem mass spectrometry.
Analysis of endocrine disrupters and related compounds in sediments and sewage sludge using on-line turbulent flow chromatography-liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2014-06-10)
Marina Gorga, Sara Insa, Mira Petrovic, Damià Barceló
ABSTRACT
A novel fully automated method based on dual column switching using turbulent flow chromatography followed by liquid chromatography coupled to tandem mass spectrometry (TFC-LC-MS/MS) was applied for the determination of endocrine disruptors (EDCs) and related compounds in sediment and sewage sludge samples. This method allows the unequivocal identification and quantification of the most relevant environmental EDCs such as natural and synthetic estrogens and their conjugates, antimicrobials, parabens, bisphenol A (BPA), alkylphenolic compounds, benzotriazoles, and organophosphorus flame retardants, minimizing time of analysis and alleviating matrix effects. Applying this technique, after the extraction of the target compounds by pressurized liquid extraction (PLE), sediment and sewage sludge extracts were directly injected to the chromatographic system and the analytes were concentrated into the clean-up loading column. Using six-port switching system, the analytes were transferred to the analytical column for subsequent detection by MS-MS (QqQ). In order to optimize this multiplexing system, a comparative study employing six types of TurboFlow™ columns, with different chemical modifications, was performed to achieve the maximum retention of analytes and best elimination of matrix components. Using the optimized protocol low limits of quantification (LOQs) were obtained ranging from 0.0083 to 1.6 ng/g for sediment samples and from 0.10 to 125 ng/g for sewage sludge samples (except for alkylphenol monoethoxylate). The method was used to evaluate the presence and fate of target EDCs in the Ebro River which is the most important river in Spain with intensive agricultural and industrial activities in the basin that contribute to deteriorating soil and water quality.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
4-tert-Octylphenol solution, 1000 μg/mL in acetone, analytical standard
Supelco
Benzotriazole, analytical standard
Sigma-Aldrich
Estriol, meets USP testing specifications
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
Ethyl salicylate, ≥99%, FCC, FG
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Supelco
Nonylphenol, PESTANAL®, analytical standard, technical mixture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Sigma-Aldrich
Nonylphenol, technical grade, mixture of ring and chain isomers
Sigma-Aldrich
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%
Sigma-Aldrich
Benzotriazole, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl salicylate, 99%
Ethinylestradiol for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Triclosan, Pharmaceutical Secondary Standard; Certified Reference Material
Estriol, European Pharmacopoeia (EP) Reference Standard
USP
Triclosan, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Diethylstilbestrol, VETRANAL®, analytical standard
Supelco
Methanol, analytical standard