Merck
  • Cryopreservation of osteoblasts by use of a programmed freezer with a magnetic field.
Cryopreservation of osteoblasts by use of a programmed freezer with a magnetic field.
Cryo letters (2013-02-26)
H Koseki, M Kaku, T Kawata, S Kojima, H Sumi, H Shikata, M Motokawa, T Fujita, J Ohtani, K Tanne
ABSTRACT
In order to determine a suitable condition for osteoblasts cryopreservation, murine osteoblasts were freezed by programmed freezer with a magnetic field (CAS freezer). After 7 days cryopreservation at -150°, the number of survival cells immediately after thawing and the growth rate of cultured cells for 48 hours were examined. Gene and protein expression of alkaline phosphatase (ALP), osteopontin (OPN) and bone sialoprotein (BSP) were compared between cryopreserved and non-cryopreserved groups. As a result, a plunging temperature of -30°, a hold-time at -5° for 15 minutes and a 0.1 mT of magnetic field led to the largest survival and growth rate. Moreover, there was no significant difference in ALP, OPN and BSP mRNA and protein expression between cryopreserved and control groups. From these results, it was suggested that the CAS freezer is available for osteoblast cryopreservation and bone tissue banking can be established in the future.
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture