Merck
  • Simultaneous stereoselective analysis of naftopidil and O-desmethyl naftopidil enantiomers in rat feces using an online column-switching high-performance liquid chromatography method.
Simultaneous stereoselective analysis of naftopidil and O-desmethyl naftopidil enantiomers in rat feces using an online column-switching high-performance liquid chromatography method.
Biomedical chromatography : BMC (2014-01-15)
Yun-Ying Zhang, Xia-Wen Liu, Li-Jun Zhu, Mu Yuan
ABSTRACT
A novel online column-switching chiral high-performance liquid chromatography method was developed and validated for the simultaneous determination of naftopidil (NAF) and its O-desmethyl metabolites (DMN) enantiomers in rat feces. Direct and multiple injections of supernatant from rat feces homogenate were allowed through the column-switching system. Analyte extraction was performed on the Capcell Pak mixed-functional column by acetonitrile-phosphate buffer (pH 7.4; 10 mm; 8:92, v/v) flowing at 1 mL/min. Separation of NAF and DMN enantiomers was achieved on the Chiralpak IA column by methanol-acetonitrile-acetate buffer (pH 5.3; 5 mm; 45:33:22, v/v/v) flowing at 0.5 mL/min. The analytes were measured with a fluorescence detector at 290 nm (λ(ex)) and 340 nm (λ(em)). The validated method showed a good linearity [22.5-15,000 ng/mL for (+)-/(-)-NAF; 35-25,000 ng/mL for (+)-/(-)-DMN] and the lowest limits of quantification for NAF and DMN enantiomers were 22.5 and 35 ng/mL, respectively. Both intra- and inter-day variations were <10%. The assay was successfully applied to the fecal excretion of NAF and DMN enantiomers in rat after single oral administration of (±)-NAF. Nonstereoselective excretion of (+)- and (-)-NAF was found in feces, while stereoselective excretion of (+)- and (-)-DMN was observed with higher excretion levels of (+)-DMN, indicating that there may exist stereoselective metabolism for NAF enantiomers.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%