Merck
  • Induction of PERV antigen in porcine peripheral blood mononuclear cells by human herpesvirus 1.
Induction of PERV antigen in porcine peripheral blood mononuclear cells by human herpesvirus 1.
Xenotransplantation (2015-02-27)
Jiyeon Kim, Jung Heon Kim, Eung Soo Hwang
ABSTRACT
Xenotransplantation represents one of alternative candidates for allotransplantation due to the chronic shortage of suitable human tissues; however, many obstacles remain. Expression and release of endogenous retroviral antigens by porcine cells after transplantation may evoke adverse immune responses in human subjects. Here, we examined whether human herpesvirus 1 (HHV-1) could induce the production of porcine endogenous retrovirus (PERV) antigens in porcine peripheral blood mononuclear cells (PBMCs). Porcine PBMCs were infected with HHV-1 and examined for the production of PERV Gag protein and HHV-1 using antigen-capture ELISA and quantitative real-time polymerase chain reaction (PCR), respectively. HHV-1 infection resulted in a 1.7- to 33.2-fold induction of PERV Gag relative to mock infection controls, compared to a 2.9- to 12.9-fold induction following treatment with PMA. Expression of PERV Gag was detected in porcine PBMCs and PK-15 cells after HHV-1 infection by double immunofluorescence staining of PERV Gag and HHV-1 antigen. The viability of HHV-1-infected porcine PBMCs was significantly lower than that of mock-infected cells. The HHV-1 level in the culture supernatant increased 5.2-fold relative to controls 24-h post-infection, indicative of active replication within these cells; decreased levels of HHV-1 were detected 72-h post-infection. These results suggest that HHV-1 may be capable of infecting transplanted porcine cells, resulting in strong direct induction of PERV antigen.
Product Description
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC