Merck
  • Mediterranean essential oils as effective weapons against the West Nile vector Culex pipiens and the Echinostoma intermediate host Physella acuta: what happens around? An acute toxicity survey on non-target mayflies.
Mediterranean essential oils as effective weapons against the West Nile vector Culex pipiens and the Echinostoma intermediate host Physella acuta: what happens around? An acute toxicity survey on non-target mayflies.
Parasitology research (2015-01-08)
Giovanni Benelli, Stefano Bedini, Guido Flamini, Francesca Cosci, Pier Luigi Cioni, Smain Amira, Fatima Benchikh, Hocine Laouer, Graziano Di Giuseppe, Barbara Conti
ABSTRACT
Mosquitoes (Diptera: Culicidae) represent a threat for millions of people worldwide, since they act as vectors for important pathogens, including malaria, yellow fever, dengue and West Nile. Second to malaria as the world's most widespread parasitic disease, infection by trematodes is a devastating public health problem. In this study, we proposed two essential oils from plants cultivated in Mediterranean regions as effective chemicals against mosquitoes and freshwater snails vectors of Echinostoma trematodes. Chemical composition of essential oils from Achillea millefolium (Asteraceae) and Haplophyllum tuberculatum (Rutaceae) was investigated. Acute toxicity was evaluated against larvae of the West Nile vector Culex pipiens (Diptera: Culicidae) and the invasive freshwater snail Physella acuta (Mollusca: Physidae), an important intermediate host of many parasites, including Echinostoma revolutum (Echinostomidae). Acute toxicity of essential oils was assessed also on a non-target aquatic organism, the mayfly Cloeon dipterum (Ephemeroptera: Baetidae). Achillea millefolium and H. tuberculatum essentials oils were mainly composed by oxygenated monoterpenes (59.3 and 71.0 % of the whole oil, respectively). Chrysanthenone and borneol were the two major constituents of Achillea millefolium essential oil (24.1 and 14.2 %, respectively). Major compounds of H. tuberculatum essential oil were cis-p-menth-2-en-1-ol and trans-p-menth-2-en-1-ol (22.9 and 16.1 %, respectively). In acute toxicity assays, C. pipiens LC50 was 154.190 and 175.268 ppm for Achillea millefolium and H. tuberculatum, respectively. P. acuta LC50 was 112.911 and 73.695 ppm for Achillea millefolium and H. tuberculatum, respectively, while the same values were 198.116 and 280.265 ppm for C. dipterum. Relative median potency analysis showed that both tested essential oils were more toxic to P. acuta over C. dipterum. This research adds knowledge on plant-borne chemicals toxic against invertebrates of medical importance, allowing us to propose the tested oils as effective candidates to develop newer and safer vector control tools.
Product Description
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)