Merck
  • Formulation approaches to improving the delivery of an antiviral drug with activity against seasonal flu.
Formulation approaches to improving the delivery of an antiviral drug with activity against seasonal flu.
Pharmaceutical development and technology (2013-11-14)
Srinivasa M Sammeta, Li Wang, Shravan K Mutyam, Kathleen O'Loughlin, Carol E Green, Milton H Werner, Terence Kelly, Gita N Shankar
ABSTRACT
The main objective of the present study was to develop formulations of noscapine hydrochloride hydrate with enhanced solubility and bioavailability using co-solvent- and cyclodextrin-based approaches. Different combinations of co-solvents, which were selected on the basis of high-throughput solubility screening, were subjected to in vitro intestinal drug permeability studies conducted with Ussing chambers. Vitamin E tocopherol polyethylene glycol succinate and propylene glycol based co-solvent formulations provided the maximum permeability coefficient for the drug. Inclusion complexes of the drug were prepared using hydroxypropyl-β-cyclodextrin and sulphobutylether cyclodextrins. Pharmacokinetic studies were carried out in male Sprague-Dawley rats for the selected formulations. The relative bioavailabilities of the drug with the co-solvent- and cyclodextrin-based formulations were found to be similar.
Product Description
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.