Merck
  • Rapid determination of nine parabens and seven other environmental phenols in urine samples of German children and adults.
Rapid determination of nine parabens and seven other environmental phenols in urine samples of German children and adults.
International journal of hygiene and environmental health (2014-07-11)
Rebecca K Moos, Jürgen Angerer, Jürgen Wittsiepe, Michael Wilhelm, Thomas Brüning, Holger M Koch
ABSTRACT
We developed a fast, selective and sensitive on-line LC/LC-MS/MS method for the simultaneous determination of nine parabens and seven environmental phenols in urine. Parabens are widely used as antimicrobial preservatives. Bisphenol A, triclosan, triclocarban, 2-phenylphenol, and benzophenones are used inter alia in disinfectants, sunscreens and in polymers. Some of these substances are suspected endocrine disruptors. Limits of quantification and analytical quality criteria fully met the needs for determining exposure levels occurring in the general population. We analyzed 157 spot urine samples from the general German population (59 females, 39 males and 59 children). For the parabens, we found methyl, ethyl and n-propyl paraben with high detection rates (77-98%), followed by n-butyl (36%), iso-butyl (17%), iso-propyl (3%) and benzyl paraben (3%). We detected no pentyl and heptyl paraben. Urinary concentrations were highest for methyl paraben (median 24.5 μg/L; 95th percentile 379 μg/L) followed by ethyl (1.4 μg/L; 35.2 μg/L) and n-propyl paraben (1.2 μg/L; 68.1 μg/L). Other environmental phenols with high detection rates were BPA (95%), triclosan (45%) and benzophenone 1 and 3 (26%). For most of the parabens/environmental phenols we found higher urinary levels in females than in males or children, probably due to differences in (personal care) product use. However, high levels (in the mg/L range) were also observed in children. Exposure to the above substances is occurring worldwide. Differences between countries do seem to exist and might be caused by different product compositions or different use habits. Human metabolism data is urgently needed to extrapolate from urinary biomarker levels to doses actually taken up.
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Heptyl 4-hydroxybenzoate, AldrichCPR
Supelco
Oxybenzone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Oxybenzone, analytical standard
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Acetic acid, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Benzyl 4-hydroxybenzoate, 99%
Sigma-Aldrich
2,2′-Dihydroxy-4-methoxybenzophenone, 98%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
2,4-Dihydroxybenzophenone, 99%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
USP
Oxybenzone, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material