Merck
  • Chemical profiling and quantification of Gua-Lou-Gui-Zhi decoction by high performance liquid chromatography/quadrupole-time-of-flight mass spectrometry and ultra-performance liquid chromatography/triple quadrupole mass spectrometry.
Chemical profiling and quantification of Gua-Lou-Gui-Zhi decoction by high performance liquid chromatography/quadrupole-time-of-flight mass spectrometry and ultra-performance liquid chromatography/triple quadrupole mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-25)
Wen Xu, Mingqing Huang, Huang Li, Xianwen Chen, Yuqin Zhang, Jie Liu, Wei Xu, Kedan Chu, Lidian Chen
ABSTRACT
Gua-Lou-Gui-Zhi decoction (GLGZD) is a classical formula of traditional Chinese medicine, which has been commonly used to treat dysfunction after stroke, epilepsy and spinal cord injury. In this study, a systematic method was established for chemical profiling and quantification analysis of the major constituents in GLGZD. For qualitative analysis, a method of high performance liquid chromatography/quadrupole time-of-flight mass spectrometry (Q-TOF MS) was developed. 106 compounds, including monoterpene glycosides, galloyl glucoses, phenolic acids, flavonoids, gingerols and triterpene saponins were identified or tentatively presumed by comparison with reference standards or literature data. According to the qualitative results, a new quantitative analysis method of ultra-performance liquid chromatography/triple quadrupole mass spectrometry (QqQ-MS) was established. 24 representative compounds were simultaneously detected in 10 batches of GLGZD samples in 7.5 min. The calibration curves for all analytes showed good linearity (r>0.9959) within the test ranges. The LODs and the LOQs were less than 30.6 and 70.9 ng/mL, respectively. The RSDs of intra- and inter-day precision, repeatability and stability were below 3.64%, 4.85%, 4.84% and 3.87%, respectively. The overall recoveries ranged from 94.94% to 103.66%, with the RSDs within 5.12%. This study established a high sensitive and efficient method for the integrating quality control, including identification and quantification of Chinese medicinal preparation.
Product Description
Supelco
Naringin, analytical standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Neochlorogenic acid, from Lonicera japonica, ≥98.0% (HPLC)
Sigma-Aldrich
Licochalcone A, ≥96.0% (HPLC)
Sigma-Aldrich
Naringin, ≥90% (HPLC), from citrus fruit
Sigma-Aldrich
Paeoniflorin, ≥98% (HPLC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Naringin, ≥95% (HPLC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Taxifolin, analytical standard
Supelco
Paeoniflorin, analytical standard
Naringin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%