Merck
  • Spectroscopic studies of 1,4-dimethoxy-2,3-dimethylanthracene-9,10-dione on plasmonic silver nanoparticles.
Spectroscopic studies of 1,4-dimethoxy-2,3-dimethylanthracene-9,10-dione on plasmonic silver nanoparticles.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2014-06-29)
S R Kavitha, M Umadevi, P Vanelle, T Terme, O Khoumeri, B Sridhar
ABSTRACT
Silver nanoparticles (Ag NPs) of different sizes from 7nm to 22nm have been prepared by simple Dirk and Charles chemical method and characterized using UV-vis spectroscopy and high resolution transmission electron microscopy (HRTEM). Fluorescence quenching of 1,4-dimethoxy-2,3-dimethylanthracene-9,10-dione (DMDMAD) by silver nanoparticles has been investigated by fluorescence spectroscopy to understand the role of quenching mechanism. Furthermore, the intensity of DMDMAD fluorescence emission peak decreases with decrease in the size of the Ag NPs. The fluorescence quenching rate constant and association constant for above system were determined using Stern-Volmer and Benesi-Hildebrand plots. The mechanism of DMDMAD fluorescence quenched by Ag NPs was discussed according to the Stern-Volmer equation. It has been observed that the quenching due to Ag NPs proceeds via dynamic quenching process. The distance between DMDMAD (donor) to Ag NPs (acceptor) and the critical energy transfer distance were estimated based on the Förster Resonance Energy Transfer (FRET) theory.
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
VenPure® SF, powder
Supelco
Rhodamine 6G, analytical standard
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%