Merck
  • Identification of noninvasive biomarkers for nephrotoxicity using HK-2 human kidney epithelial cells.
Identification of noninvasive biomarkers for nephrotoxicity using HK-2 human kidney epithelial cells.
Toxicological sciences : an official journal of the Society of Toxicology (2014-07-02)
Sun Young Kim, So-Jung Sohn, A Jin Won, Hyung Sik Kim, Aree Moon
ABSTRACT
The kidney is an important site of xenobiotic-induced toxicity. Because the traditional markers of renal injury indicate only severe renal damage, new biomarkers are needed for a more sensitive and reliable evaluation of renal toxicity. This study was designed to identify in vitro noninvasive biomarkers for efficient assessment of nephrotoxicity by using cisplatin as a model of nephrotoxic compounds. To this end, a comparative proteomic analysis of conditioned media from HK-2 human kidney epithelial cells treated with cisplatin was performed. Here, we identified pyruvate kinase M1/M2 isoform M2 (PKM2) and eukaryotic translation elongation factor 1 gamma (EF-1γ) as potential biomarker candidates for evaluation of nephrotoxicity. PKM2 and EF-1γ were increased by cisplatin in a kidney cell-specific manner, most likely due to cisplatin-induced apoptosis. The increase of PKM2 and EF-1γ levels in conditioned media was also observed in the presence of other nephrotoxic agents with different cytotoxic mechanisms such as CdCl2, HgCl2, and cyclosporine A. Rats treated with cisplatin, CdCl2, or HgCl2 presented increased levels of PKM2 and EF-1γ in the urine and kidney tissue. Taken together, this study identified two noninvasive biomarker candidates, PKM2 and EF-1γ, by comparative proteomic analysis. These new biomarkers may offer an alternative to traditional renal markers for efficient evaluation of nephrotoxicity.
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mercury(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Mercury(II) chloride, ACS reagent, ≥99.5%
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cadmium chloride, technical grade