Merck
  • Prophylactic use of ketorolac tromethamine in cataract surgery: a randomized trial.
Prophylactic use of ketorolac tromethamine in cataract surgery: a randomized trial.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics (2014-04-17)
Flavia G Ticly, Rodrigo P C Lira, Fernando R Zanetti, Maria Cecília Machado, Gustavo B Rodrigues, Carlos Eduardo L Arieta
ABSTRACT
To determine the efficacy of topical ketorolac tromethamine in preventing cystoid macular edema (CME) after uncomplicated cataract surgery. This single-center, prospective, double-masked, randomized clinical trial consisted of 81 patients who were scheduled for cataract surgery. Patients were randomized to receive hypromellose/dextran 70 as a placebo (n=44) or ketorolac tromethamine 0.4% (n=37) as an adjuvant therapy. These eye drops were administered 4 times daily (QID) for 3 days before surgery and 5 weeks postoperatively. All patients received prednisolone acetate 1% QID during the same period as basal/standard anti-inflammatory therapy. The primary outcome was the incidence of angiographic CME 5 weeks after surgery. The secondary outcomes were mean change in best-corrected visual acuity (BCVA) [Early Treatment Diabetic Retinopathy study (ETDRS)], clinical CME incidence, intraocular pressure, and retinal thickness measured using optical coherence tomography (OCT). In the placebo group, 2/44 (4.5%) patients and in the ketorolac group, 2/37 (5.4%) patients presented with angiographic CME (P=0.624). The mean change in postoperative BCVA was 32±15 letters in the placebo group and 26±16 letters in the ketorolac group (P=0.07). There were no statistically significant between-group differences in the mean central subfield thickness (P=0.679), minimal central thickness (P=0.352), or central macular volume (P=0.729). There was no difference between ketorolac tromethamine and a placebo with regard to BCVA results or prevention of CME after uncomplicated cataract surgery.
Sigma-Aldrich
Prednisolone 21-acetate, ≥97%
Sigma-Aldrich
Prednisolone, ≥99%
Supelco
Prednisolone, VETRANAL®, analytical standard
Supelco
Prednisolone, Pharmaceutical Secondary Standard; Certified Reference Material
Prednisolone for system suitability, European Pharmacopoeia (EP) Reference Standard
Prednisolone, European Pharmacopoeia (EP) Reference Standard
Supelco
Ketorolac Tromethamine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ketorolac Tromethamine, United States Pharmacopeia (USP) Reference Standard
Ketorolac trometamol for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Prednisolone, United States Pharmacopeia (USP) Reference Standard
Prednisolone acetate for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Ketorolac tris salt, ≥99%, crystalline
Prednisolone for peak identification, European Pharmacopoeia (EP) Reference Standard
Prednisolone acetate, European Pharmacopoeia (EP) Reference Standard
USP
Prednisolone acetate, United States Pharmacopeia (USP) Reference Standard
Ketorolac trometamol, European Pharmacopoeia (EP) Reference Standard