Merck
  • Identification of photosynthesis inhibitors of pelagic marine algae using 96-well plate microfractionation for enhanced throughput in effect-directed analysis.
Identification of photosynthesis inhibitors of pelagic marine algae using 96-well plate microfractionation for enhanced throughput in effect-directed analysis.
Environmental science & technology (2014-06-14)
Petra Booij, A Dick Vethaak, Pim E G Leonards, Sascha B Sjollema, Jeroen Kool, Pim de Voogt, Marja H Lamoree
ABSTRACT
Because of large-scale production and use of an increasing diversity of chemicals in modern society, estuarine and coastal waters may be contaminated with numerous substances. Some of these compounds have the potential to affect microalgae at the base of the pelagic food chain. Therefore, we identified the main chemical stressors that negatively affect the effective photosystem II efficiency (ϕPSII) in marine microalgae of the Dutch estuarine and coastal waters. An enhanced effect-directed analysis (EDA) was carried out by combining reversed-phase ultra performance liquid chromatography fractionation of extracts from passive samplers, followed by effect assessment using the pulse amplitude modulation fluorometry assay and chemical analysis of biologically active fractions using high-resolution mass spectrometry. This study focuses on a novel microfractionation technique using 96-well plates to enhance throughput in EDA, structure elucidation, and the analytical and effect confirmation of the compounds that are identified. Although there are numerous unknown compounds present in estuarine and coastal waters, our EDA study shows that atrazine, diuron, irgarol, isoproturon, terbutryn, and terbutylazine are the main contributors to the observed effect on the ϕPSII of marine microalgae.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Pyrrolidone, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Pyrrolidinone, purum, ≥98.0% (GC)
Sigma-Aldrich
2-Pyrrolidinone, ≥99%
Sigma-Aldrich
2-Pyrrolidinone, 99%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Supelco
Glycerol, analytical standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, FCC, FG