Merck
  • Investigation of the therapeutic effectiveness of active components in Sini decoction by a comprehensive GC/LC-MS based metabolomics and network pharmacology approaches.
Investigation of the therapeutic effectiveness of active components in Sini decoction by a comprehensive GC/LC-MS based metabolomics and network pharmacology approaches.
Molecular bioSystems (2014-10-16)
Si Chen, Si Wu, Wuhong Li, Xiaofei Chen, Xin Dong, Guangguo Tan, Hai Zhang, Zhanying Hong, Zhenyu Zhu, Yifeng Chai
ABSTRACT
As a classical formula, Sini decoction (SND) has been fully proved to be clinically effective in treating doxorubicin (DOX)-induced cardiomyopathy. Current chemomics and pharmacology proved that the total alkaloids (TA), total gingerols (TG), total flavones and total saponins (TFS) are the major active ingredients of Aconitum carmichaelii, Zingiber officinale and Glycyrrhiza uralensis in SND respectively. Our animal experiments in this study demonstrated that the above active ingredients (TAGFS) were more effective than formulas formed by any one or two of the three individual components and nearly the same as SND. However, very little is known about the action mechanisms of TAGFS. Thus, this study aimed to use for the first time the combination of GC/LC-MS based metabolomics and network pharmacology for solving this problem. By metabolomics, it was found that TAGFS worked by regulating six primary pathways. Then, network pharmacology was applied to search for specific targets. 17 potential cardiovascular related targets were found through molecular docking, 11 of which were identified by references, which demonstrated the therapeutic effectiveness of TAGFS using network pharmacology. Among these targets, four targets, including phosphoinositide 3-kinase gamma, insulin receptor, ornithine aminotransferase and glucokinase, were involved in the TAGFS regulated pathways. Moreover, phosphoinositide 3-kinase gamma, insulin receptor and glucokinase were proved to be targets of active components in SND. In addition, our data indicated TA as the principal ingredient in the SND formula, whereas TG and TFS served as adjuvant ingredients. We therefore suggest that dissecting the mode of action of clinically effective formulae with the combination use of metabolomics and network pharmacology may be a good strategy.
Product Description
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Supelco
Acetone, analytical standard
Supelco
N-Methyl-bis(trifluoroacetamide), for GC derivatization, LiChropur, ≥99.0% (GC)
Supelco
N-Methyl-bis(trifluoroacetamide), for GC derivatization, LiChropur, ≥97.0% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methoxyamine hydrochloride, 98%
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Heptane, analytical standard
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Heptane, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥96%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Heptane, HPLC Plus, for HPLC, GC, and residue analysis, 99%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Heptane, puriss. p.a., reag. Ph. Eur., ≥99% n-heptane basis (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%