Merck
  • Expansion of toll-like receptor 9-expressing B cells in active systemic lupus erythematosus: implications for the induction and maintenance of the autoimmune process.
Expansion of toll-like receptor 9-expressing B cells in active systemic lupus erythematosus: implications for the induction and maintenance of the autoimmune process.
Arthritis and rheumatism (2006-11-01)
Eva D Papadimitraki, Christianna Choulaki, Eleni Koutala, George Bertsias, Christos Tsatsanis, Irini Gergianaki, Amalia Raptopoulou, Heraklis D Kritikos, Clio Mamalaki, Prodromos Sidiropoulos, Dimitrios T Boumpas
ABSTRACT
Toll-like receptors (TLRs) are pattern-associated receptors in innate immunity that may be involved in the recognition of self antigens and the production of pathogenic autoantibodies. This study was undertaken to examine the expression and function of various TLRs in subpopulations of peripheral blood mononuclear cells (PBMCs) of patients with systemic lupus erythematosus (SLE). The expression of TLRs in PBMCs from 50 SLE patients with active disease (SLE Disease Activity Index [SLEDAI] score >or=8; n = 26) or inactive disease (SLEDAI score <8; n = 24) and 20 healthy controls was studied by flow cytometry. TLR expression was assessed on various subpopulations of PBMCs (TLR-2 and TLR-4 by membrane staining; TLR-3 and TLR-9 by intracellular staining). TLR function was accessed by stimulating PBMCs with specific ligands. The proportion of B cells and monocytes expressing TLR-9 was higher among patients with active SLE (mean +/- SD 49.5 +/- 24.4% and 30.7 +/- 24.1%, respectively) than among patients with inactive disease (22.8 +/- 19.6% and 14.3 +/- 8.4%, respectively; P = 0.02 and P = 0.03). Among B cells, the proportion of plasma cells and memory B cells expressing TLR-9 was increased in patients with active SLE. Increased percentages of TLR-9-expressing B cells correlated with the presence of anti-double-stranded DNA antibodies (P = 0.007). Treatment with serum from patients with active disease increased the percentage of TLR-9-expressing plasma cells in serum from healthy controls. Enhanced induction of HLA-DR after TLR-9 stimulation was documented in B cells from patients with active disease. In patients with active SLE, the proportion of peripheral blood memory B cells and plasma cells expressing TLR-9 is increased. Endogenous nucleic acids released during apoptotic cell death may stimulate B cells via TLR-9 and contribute to SLE pathogenesis.
Sigma-Aldrich
Polyinosinic–polycytidylic acid sodium salt, γ-irradiated
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
Sigma-Aldrich
Sodium pyruvate solution, 100 mM, sterile-filtered, BioReagent, suitable for cell culture