Merck
  • Differential induction of pro- and anti-inflammatory cytokines in whole blood by bacteria: effects of antibiotic treatment.
Differential induction of pro- and anti-inflammatory cytokines in whole blood by bacteria: effects of antibiotic treatment.
Antimicrobial agents and chemotherapy (1997-07-01)
J T Frieling, J A Mulder, T Hendriks, J H Curfs, C J van der Linden, R W Sauerwein
ABSTRACT
The in vitro production of interleukin-1beta (IL-1beta), IL-6, and the IL-1 receptor antagonist (IL-1ra) in whole blood upon stimulation with different bacterial strains was measured to study the possible relationship between disease severity and the cytokine-inducing capacities of these strains. Escherichia coli, Neisseria meningitidis, Neisseria gonorrhoeae, Bacteroides fragilis, Capnocytophaga canimorsus, Staphylococcus aureus, Enterococcus faecalis, Streptococcus pneumoniae, and Streptococcus pyogenes induced the cytokines IL-1beta, IL-6, and IL-1ra. Gram-negative bacteria induced significantly higher levels of proinflammatory cytokine production than gram-positive bacteria. These differences were less pronounced for the anti-inflammatory cytokine IL-1ra. In addition, blood was stimulated with E. coli killed by different antibiotics to study the effect of the antibiotics on the cytokine-inducing capacity of the bacterial culture. E. coli treated with cefuroxime and gentamicin induced higher levels of IL-1beta and IL-6 production but levels of IL-1ra production similar to that of heat-killed E. coli. In contrast, ciprofloxacin- and imipenem-cilastatin-mediated killing showed a decreased or similar level of induction of cytokine production as compared to that by heat-killed E. coli; polymyxin B decreased the level of production of the cytokines.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by trichloroacetic acid extraction
Sigma-Aldrich
Streptavidin−Peroxidase from Streptomyces avidinii, lyophilized powder