Merck
  • Acute blood pressure effects and chronic hypotensive action of calcimimetics in uremic rats.
Acute blood pressure effects and chronic hypotensive action of calcimimetics in uremic rats.
Journal of the American Society of Nephrology : JASN (2006-01-20)
Tobias Odenwald, Kumiko Nakagawa, Charlotte Hadtstein, Frank Roesch, Peter Gohlke, Eberhard Ritz, Franz Schaefer, Claus Peter Schmitt
ABSTRACT
A previous study in subtotally nephrectomized (SNX) rats suggested beneficial effects of the calcimimetic R-568 beyond the control of mineral metabolism. This study analyzed potential blood pressure (BP)-lowering effects of R-568. Male Sprague-Dawley rats received two-stage subtotal nephrectomy or sham operation. Telemetry devices were inserted into the abdominal aorta, and BP was measured every 5 min. R-568 (20 mg/kg per d) or solvent was infused for 4 wk followed by once-daily subcutaneous injections for 2 wk. Total body sodium was measured by neutron activation analysis. The uremia-induced increase of mean arterial pressure from baseline to day 42 in SNX solvent rats (103+/-5 to 128+/-14 mmHg, P=0.006) was attenuated by R-568 (104+/-5 to 111+/-8 mmHg; P<0.0001 for difference of slopes). The circadian rhythm was abrogated in SNX rats and not restored by R-568. In sham-operated rats, R-568 had only a minor transient antihypertensive effect. R-568 injection induced a transient rise of mean arterial pressure by 23+/-4 and 26+/-10 mmHg in sham and SNX rats but only by 9+/-3 and 10+/-5 mmHg in solvent-treated rats (P<0.01 versus baseline and solvent versus R-568). Plasma angiotensin-converting enzyme activity and aldosterone levels were similar; food intake and physical activity did not differ throughout the study. In healthy rats, total body sodium was higher after 14 d of R-568 compared with solvent infusion (37.1+/-4 versus 32.5+/-1.4 mmol/kg; P=0.01). The calcimimetic R-568 causes an initial BP increase in sham-operated and uremic rats, which in uremic rats is followed by a marked and sustained antihypertensive effect.
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined