Merck
  • BH-protocadherin-c, a member of the cadherin superfamily, interacts with protein phosphatase 1 alpha through its intracellular domain.
BH-protocadherin-c, a member of the cadherin superfamily, interacts with protein phosphatase 1 alpha through its intracellular domain.
FEBS letters (1999-11-26)
K Yoshida, M Watanabe, H Kato, A Dutta, S Sugano
ABSTRACT
Using a yeast two-hybrid system, we isolated eight cDNA clones which interacted with BH-protocadherin-c (BH-Pcdh-c) from the human brain cDNA library. One clone encoded protein phosphatase type I isoform alpha (PP1alpha) and another two PP1alpha2. PP1alpha was co-immunoprecipitated from the extract of a gastric adenocarcinoma cell line MKN-28 with anti-BH-Pcdh-c antibody. PP1alpha activity towards glycogen phosphorylase was inhibited by the intracellular domain of BH-Pcdh-c. Inhibition of the phosphatase required more than the minimal domain of BH-Pcdh-c which could associate with PP1alpha. In situ hybridization revealed that BH-Pcdh-c mRNA was predominantly expressed in cerebral cortex neurons in the adult mouse brain.