Merck
  • Rapid method for determination of carbonyl groups in lignin compounds by headspace gas chromatography.
Rapid method for determination of carbonyl groups in lignin compounds by headspace gas chromatography.
Journal of chromatography. A (2015-06-09)
Jing Li, Hui-Chao Hu, Xin-Sheng Chai
ABSTRACT
The paper reports on a novel method for rapid determination of carbonyl in lignins by headspace gas chromatography (HS-GC). The method involves the quantitative carbonyl reduction for aldehydes in 2min at room temperature or for acetones in 30min at 80°C by sodium borohydride solution in a closed headspace sample vial. After the reaction, the solution was acidified by injecting sulfuric acid solution and the hydrogen released to the headspace was determined by GC using thermal-conductivity detector. The results showed that with the addition of SiO2 powder, the reduction reaction of carbonyl groups can be greatly facilitated. The method has a good measurement precision (RSD<7.74%) and accuracy (relative error <10% compared with a reference method) in the carbonyl quantification. It is suitable to be used for rapid determination of carbonyl content in lignin and related materials.
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%