Merck
  • ZNF139 promotes tumor metastasis by increasing migration and invasion in human gastric cancer cells.
ZNF139 promotes tumor metastasis by increasing migration and invasion in human gastric cancer cells.
Neoplasma (2014-05-16)
Y Li, B B Tan, Q Zhao, L Q Fan, D Wang, Y Liu
ABSTRACT
Zinc finger protein 139(ZNF139), a member of zinc finger protein family, is a transcription factor. Our previous research showed ZNF139 was overexpressed in gastric cancer cells. The purpose of present study is to explore impact and mechanism of ZNF139 on metastasis by regulating invasive ability of gastric cancer cells. Quantitative RT-PCR(QRT-PCR) and Western blot were applied for detection of ZNF139 expression in gastric cancer tissues, adjacent cancer tissues, metastatic lymph nodes, gastric cancer cell lines SGC7901 and BGC823 and gastric epithelial cell line GES-1; siRNA specific to ZNF139 was synthesized and then transfected into gastric cancer cell line BGC823; wound healing assay and Transwell assay were used to observe impact of ZNF139-siRNA after being transfected into BGC823 on its invasion and migration; changes in expression of invasion and migration-related genes MMP-2, MMP-9, ICAM-1 and TIMP1 were detected before and after transfection. Gelatin zymogrphy assay were applied to determine the MMP activities. Statistical analysis was based on the SPSS11.5 software.Expression of ZNF139 in gastric adenocarcinoma tissues and cells was significantly higher than the expression in the adjacent cancer tissues, but lower than the expression in the metastatic lymph nodes; ZNF139 expression was present in gastric cancer cell lines, and the expression level was higher than that in normal gastric epithelial cells lines. ZNF139-siRNA significantly inhibited the invasion and migration activity of gastric cancer cell line BGC823. 48h after ZNF139-siRNA was transfected into gastric cancer cell line BGC823, expression and activity of invasion-related genes MMP-2, MMP-9, ICAM-1 mRNA and protein were significantly inhibited, while expressions of TIMP-1 mRNA and protein were significantly increased. At the same time, the gelatinase activities of MMP2 and MMP9 were decreased by ZNF139 interference.ZNF139 was overexpressed in gastric cancer cells, and the expression was further enhanced in the metastasis process. Knocking down ZNF139 expression in gastric cancer cells could effectively reduce gastric cancer cell invasion and migration ability, and this process might play a role by regulating MMP-TIMP balance.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Fluorescein, European Pharmacopoeia (EP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard