Merck
  • The Freud-1/CC2D1A family: transcriptional regulators implicated in mental retardation.
The Freud-1/CC2D1A family: transcriptional regulators implicated in mental retardation.
Journal of neuroscience research (2007-03-31)
Anastasia Rogaeva, Kimberly Galaraga, Paul R Albert
ABSTRACT
The CC2D1A gene family consists of two homologous genes, Freud-1/CC2D1A and Freud-2/CC2D1B, that share conserved domains, including several DM14 domains that are specific to this protein family, a C-terminal helix-loop-helix domain, and a C2 calcium-dependent phospholipid binding domain. Although the function of Freud-2 is unknown, Freud-1 has been shown to function as a transcriptional repressor of the serotonin-1A receptor gene that binds to a novel DNA element (FRE, 5'-repressor element). The DNA binding and repressor activities of Freud-1 are inhibited by calcium-calmodulin-dependent protein kinase. Recently, a deletion in the CC2D1A gene has been linked to nonsyndromic mental retardation. This deletion results in the truncation of the helix-loop-helix DNA binding and the C2 domains, crucial for Freud-1 repressor activity, and hence is predicted to generate an inactive or weakly dominant negative protein. The possible mechanisms by which inactivation of Freud-1 could lead to abnormal cortical development and cognitive impairment and the potential roles of Freud-1 gene targets are discussed.