Merck
  • Knockdown of ZFX suppresses renal carcinoma cell growth and induces apoptosis.
Knockdown of ZFX suppresses renal carcinoma cell growth and induces apoptosis.
Cancer genetics (2014-12-03)
Qiang Fang, Wei-hua Fu, Jing Yang, Xing Li, Zhan-song Zhou, Zhi-weng Chen, Jin-hong Pan
ABSTRACT
The ZFX (zinc finger protein, X-linked) gene located on the human X chromosome controls the self-renewal of embryonic and hematopoietic stem cells as a transcriptional regulator. Recently, studies have affirmed that ZFX is associated with several human cancers, including lymphoma, laryngeal squamous cell carcinoma, prostate cancer, and liver cancer, which suggests ZFX as a potential therapeutic target in cancer. However, the functional role of ZFX in human renal cancer remains unclear. Herein, we detected the expression of ZFX in 42 patients with renal cancer and found the expression of ZFX was specifically upregulated in cancer tissues at the mRNA and protein levels. Moreover, we employed lentivirus-mediated short hairpin RNA (shRNA) to knock down ZFX expression in two human renal cell carcinoma cell lines, 786-0 and ACHN. Functional analysis indicated that ZFX silencing significantly inhibited renal cell carcinoma cell proliferation and cell cycle progression, probably because of suppression of CDK4 and cyclin D1, and induced apoptosis via activation of Bax, Caspase 3, and PUMA in a p53-dependent manner. Our findings suggest that knockdown of ZFX by shRNA may be a potential therapeutic approach for the treatment of renal cancer.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Propidium iodide solution
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O