Merck
  • DJBP: a novel DJ-1-binding protein, negatively regulates the androgen receptor by recruiting histone deacetylase complex, and DJ-1 antagonizes this inhibition by abrogation of this complex.
DJBP: a novel DJ-1-binding protein, negatively regulates the androgen receptor by recruiting histone deacetylase complex, and DJ-1 antagonizes this inhibition by abrogation of this complex.
Molecular cancer research : MCR (2003-03-04)
Takeshi Niki, Kazuko Takahashi-Niki, Takahiro Taira, Sanae M M Iguchi-Ariga, Hiroyoshi Ariga
ABSTRACT
DJ-1 was identified by us as a novel oncogene that transforms mouse NIH3T3 cells in cooperation with ras. We later identified PIAS (protein inhibitor of activated STAT)xalpha as a DJ-1-binding protein, and found that DJ-1 restored androgen receptor (AR) transcription activity that was repressed by PIASxalpha. To further characterize the function of DJ-1, we cloned cDNA encoding a novel DJ-1-binding protein, DJBP, by a yeast two-hybrid system. DJBP mRNA was found to be specifically expressed in the testis. In addition to the binding of DJBP to the COOH-terminal region of DJ-1, DJBP was also found to bind in vitro and in vivo to the DNA-binding domain of the AR in a testosterone-dependent manner and to be colocalized with DJ-1 or AR in the nucleus. Furthermore, a co-immunoprecipitation assay showed that the formation of a ternary complex between DJ-1, DJBP, and AR occurred in cells in which DJ-1 bound to the AR via DJBP. It was found that DJBP repressed a testosterone-dependent AR transactivation activity in monkey Cos1 cells by recruiting histone deacetylase (HDAC) complex, including HDAC1 and mSin3, and that DJ-1 partially restored its repressed activity by abrogating DJBP-HDAC complex. These results suggest that AR is positively regulated by DJ-1, which antagonizes the function of negative regulators, including DJBP.