Merck
  • Calcium-sensing receptor-mediated osteogenic and early-stage neurogenic differentiation in umbilical cord matrix mesenchymal stem cells from a large animal model.
Calcium-sensing receptor-mediated osteogenic and early-stage neurogenic differentiation in umbilical cord matrix mesenchymal stem cells from a large animal model.
PloS one (2014-11-08)
Nicola Antonio Martino, Stephan Joel Reshkin, Elena Ciani, Maria Elena Dell'Aquila
ABSTRACT
Umbilical cord matrix mesenchymal stem cells (UCM-MSCs) present a wide range of potential therapeutical applications. The extracellular calcium-sensing receptor (CaSR) regulates physiological and pathological processes. We investigated, in a large animal model, the involvement of CaSR in triggering osteogenic and neurogenic differentiation of two size-sieved UCM-MSC lines, by using AMG641, a novel potent research calcimimetic acting as CaSR agonist. Large (>8 µm in diameter) and small (<8 µm) equine UCM-MSC lines were cultured in medium with high calcium (Ca2+) concentration ([Ca2+]o; 2.87 mM) and dose-response effects of AMG641 (0.01 to 3µM) on cell proliferation were evaluated. Both cell lines were then cultured in osteogenic or neurogenic differentiation medium containing: 1) low [Ca2+]o (0.37 mM); 2) high [Ca2+]o (2.87 mM); 3) AMG641 (0.05, 0.1 or 1 µM) with high [Ca2+]o and 4) the CaSR antagonist NPS2390 (10 mM for 30 min) followed by incubation with AMG641 in high [Ca2+]o. Expression of osteogenic or neurogenic differentiation biomarkers was compared among groups. In both cell lines, AMG641 dose-dependently increased cell proliferation (up to P<0.001). Osteogenic molecular markers expression was differentially regulated by AMG641, with stimulatory (OPN up-regulation) in large or inhibitory (RUNX2 and OPN down-regulation) effects in small cells, respectively. AMG641 significantly increased alkaline phosphatase activity and calcium phosphate deposition in both cell lines. Following treatment with AMG641 during osteogenic differentiation, in both cell lines CaSR expression was inversely related to that of osteogenic markers and inhibition of CaSR by NPS2390 blocked AMG641-dependent responses. Early-stage neurogenic differentiation was promoted/triggered by AMG641 in both cell lines, as Nestin and CaSR mRNA transcription up-regulation were observed. Calcium- and AMG641-induced CaSR stimulation promoted in vitro proliferation and osteogenic and early-stage neurogenic differentiation of UCM-MSCs. CaSR activation may play a fundamental role in selecting specific differentiation checkpoints of these two differentiation routes, as related to cell commitment status.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water