Merck
  • Titer on chip: new analytical tool for influenza vaccine potency determination.
Titer on chip: new analytical tool for influenza vaccine potency determination.
PloS one (2014-10-21)
Laura R Kuck, Michelle Sorensen, Erin Matthews, Indresh Srivastava, Manon M J Cox, Kathy L Rowlen
ABSTRACT
Titer on Chip (Flu-ToC) is a new technique for quantification of influenza hemagglutinin (HA) concentration. In order to evaluate the potential of this new technique, a comparison of Flu-ToC to more conventional methods was conducted using recombinant HA produced in a baculovirus expression system as a test case. Samples from current vaccine strains were collected from four different steps in the manufacturing process. A total of 19 samples were analysed by Flu-ToC (blinded), single radial immunodiffusion (SRID), an enzyme-linked immunosorbent assay (ELISA), and the purity adjusted bicinchoninic acid assay (paBCA). The results indicated reasonable linear correlation between Flu-ToC and SRID, ELISA, and paBCA, with regression slopes of log-log plots being 0.91, 1.03, and 0.91, respectively. The average ratio for HA content measured by Flu-ToC relative to SRID, ELISA, and paBCA was 83%, 147%, and 81%, respectively; indicating nearly equivalent potency determination for Flu-ToC relative to SRID and paBCA. These results, combined with demonstrated multiplexed analysis of all components within a quadrivalent formulation and robust response to HA strains over a wide time period, support the conclusion that Flu-ToC can be used as a reliable and time-saving alternative potency assay for influenza vaccines.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Brilliant Blue R, Dye content ~50 %, Technical grade
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Supelco
Glycerol, analytical standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard