Merck
  • Store-operated Ca2+ entry does not control proliferation in primary cultures of human metastatic renal cellular carcinoma.
Store-operated Ca2+ entry does not control proliferation in primary cultures of human metastatic renal cellular carcinoma.
BioMed research international (2014-08-16)
Silvia Dragoni, Ilaria Turin, Umberto Laforenza, Duilio Michele Potenza, Cinzia Bottino, Toma N Glasnov, Martina Prestia, Federica Ferulli, Anna Saitta, Alessandra Mosca, Germano Guerra, Vittorio Rosti, Ombretta Luinetti, Carlo Ganini, Camillo Porta, Paolo Pedrazzoli, Franco Tanzi, Daniela Montagna, Francesco Moccia
ABSTRACT
Store-operated Ca(2+) entry (SOCE) is activated following depletion of the inositol-1,4,5-trisphosphate (InsP3)-sensitive Ca(2+) pool to regulate proliferation in immortalized cell lines established from either primary or metastatic lesions. The molecular nature of SOCE may involve both Stim1, which senses Ca(2+) levels within the endoplasmic reticulum (ER) Ca(2+) reservoir, and a number of a Ca(2+)-permeable channels on the plasma membrane, including Orai1, Orai3, and members of the canonical transient receptor (TRPC1-7) family of ion channels. The present study was undertaken to assess whether SOCE is expressed and controls proliferation in primary cultures isolated from secondary lesions of heavily pretreated metastatic renal cell carcinoma (mRCC) patients. SOCE was induced following pharmacological depletion of the ER Ca(2+) store, but not by InsP3-dependent Ca(2+) release. Metastatic RCC cells express Stim1-2, Orai1-3, and TRPC1-7 transcripts and proteins. In these cells, SOCE was insensitive to BTP-2, 10 µM Gd(3+) and Pyr6, while it was inhibited by 100 µM Gd(3+), 2-APB, and carboxyamidotriazole (CAI). Neither Gd(3+) nor 2-APB or CAI impaired mRCC cell proliferation. Consistently, no detectable Ca(2+) signal was elicited by growth factor stimulation. Therefore, a functional SOCE is expressed but does not control proliferation of mRCC cells isolated from patients resistant to multikinase inhibitors.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethidium bromide, BioReagent, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Ethidium bromide solution, BioReagent, 10 mg/mL in H2O
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Ethidium bromide solution, BioReagent, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, ~1% in H2O
Sigma-Aldrich
Prestained Molecular Weight Marker, mol wt 26,600-180,000 Da