Merck
  • Nasal allergen deposition leads to conjunctival mast cell degranulation in allergic rhinoconjunctivitis.
Nasal allergen deposition leads to conjunctival mast cell degranulation in allergic rhinoconjunctivitis.
American journal of rhinology & allergy (2014-09-10)
Ina Callebaut, Annick De Vries, Brecht Steelant, Valérie Hox, Sonja Bobic, Laura Van Gerven, Jan L Ceuppens, Peter W Hellings
ABSTRACT
The naso-ocular interaction in allergic rhinoconjunctivitis is well recognized from epidemiological, clinical, and experimental observations. The precise mechanisms remain incompletely understood. A new mouse model of allergic rhinoconjunctivitis was used to investigate the contribution of mast cells and trigeminal ganglia activation to conjunctival (conj.) inflammation after nasal allergen provocation. Sensitized mice were exposed to ovalbumin (OVA) via the nose and/or conjunctiva, and conj. homogenates were analyzed for histamine and substance P (using ELISA) and by eosinophil peroxidase (EPO) and beta-hexosaminidase assays. The conj. effects of nasal allergen deposition were compared with those induced by the mast cell activator C48/80 and with pretreatment of the mast cell stabilizer ketotifen or the transient receptor potential channel receptor (TRP) agonist capsaicin. Protachykinin 1 (TAC1) expression was quantified in the trigeminal ganglia using real time polymerase chain reaction. At 1 hour after nasal application of OVA, increased conj. levels of beta-hexosaminidase (0.68 ± 0.03 nm versus 0.56 ± 0.02 nm; p = 0.02), histamine (751.1 ± 52.17 ng/mL versus 546.3 ± 76.91 ng/mL; p = 0.05), and EPO (0.66 ± 0.09 nm versus 0.37 ± 0.03 nm; p = 0.02) were detected compared with saline. Higher levels of TAC1 expression were found in the trigeminal ganglia at 24 hours after OVA application (1326 ± 255 versus 687.5 ± 90.77 TAC1/beta-actin; p = 0.04). Nasal challenge with C48/80 increased substance P and beta-hexosaminidase levels in the conjunctiva, as well as TAC1 expression. Pretreatment with ketotifen resulted in lower levels of substance P as well as TAC1 expression. Destruction of sensory nerves in the nose by capsaicin reduced the OVA-induced conj. levels of substance P, histamine, and beta-hexosaminidase. Nasal allergen deposition in sensitized mice induced trigeminal TAC1 expression and conj. mast cell degranulation. These data represent a significant step forward in understanding the close interaction between nasal and conj. inflammation in allergy.
Sigma-Aldrich
N-Vanillylnonanamide, ≥97%, powder
Pricing and availability is not currently available.
Supelco
N-Vanillylnonanamide, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Compound 48/80
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
Pricing and availability is not currently available.