Merck
  • Lipid phosphate phosphatase-1 expression in cancer cells attenuates tumor growth and metastasis in mice.
Lipid phosphate phosphatase-1 expression in cancer cells attenuates tumor growth and metastasis in mice.
Journal of lipid research (2014-09-12)
Xiaoyun Tang, Matthew G K Benesch, Jay Dewald, Yuan Y Zhao, Neeraj Patwardhan, Webster L Santos, Jonathan M Curtis, Todd P W McMullen, David N Brindley
ABSTRACT
Lipid phosphate phosphatase-1 (LPP1) degrades lysophosphatidate (LPA) and attenuates receptor-mediated signaling. LPP1 expression is low in many cancer cells and tumors compared with normal tissues. It was hypothesized from studies with cultured cells that increasing LPP1 activity would decrease tumor growth and metastasis. This hypothesis has never been tested in vivo. To do this, we inducibly expressed LPP1 or a catalytically inactive mutant in cancer cells. Expressing active LPP1 increased extracellular LPA degradation by 5-fold. It also decreased the stimulation of Ca(2+) transients by LPA, a nondephosphorylatable LPA1/2 receptor agonist and a protease-activated receptor-1 peptide. The latter results demonstrate that LPP1 has effects downstream of receptor activation. Decreased Ca(2+) mobilization and Rho activation contributed to the effects of LPP1 in attenuating the LPA-induced migration of MDA-MB-231 breast cancer cells and their growth in 3D culture. Increasing LPP1 expression in breast and thyroid cancer cells decreased tumor growth and the metastasis by up to 80% compared with expression of inactive LPP1 or green fluorescent protein in syngeneic and xenograft mouse models. The present work demonstrates for the first time that increasing the LPP1 activity in three lines of aggressive cancer cells decreases their abilities to produce tumors and metastases in mice.
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Probenecid
Avanti
Sphingosine-1-Phosphate (d18:1), Avanti Research - A Croda Brand
Sigma-Aldrich
Apo-Direct TUNEL Assay Kit, The APO-DIRECT Kit is a single-step staining method for labeling DNA breaks to detect apoptotic cells by flow cytometry.
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
U-73122 hydrate, powder