Merck
  • Nrf2-inducing anti-oxidation stress response in the rat liver--new beneficial effect of lansoprazole.
Nrf2-inducing anti-oxidation stress response in the rat liver--new beneficial effect of lansoprazole.
PloS one (2014-05-23)
Yasunobu Yamashita, Takashi Ueyama, Toshio Nishi, Yuta Yamamoto, Akatsuki Kawakoshi, Shogo Sunami, Mikitaka Iguchi, Hideyuki Tamai, Kazuki Ueda, Takao Ito, Yoshihiro Tsuruo, Masao Ichinose
ABSTRACT
Lansoprazole is a potent anti-gastric ulcer drug that inhibits gastric proton pump activity. We identified a novel function for lansoprazole, as an inducer of anti-oxidative stress responses in the liver. Gastric administration of lansoprazole (10-100 mg/kg) to male Wistar rats produced a dose-dependent increase in hepatic mRNA levels of nuclear factor, erythroid-derived 2, -like 2 (Nrf2), a redox-sensitive transcription factor, at 3 h and Nrf2 immunoreactivity (IR) in whole hepatic lysates at 6 h. Conversely, the levels of Kelch-like ECH-associated protein (Keap1), which sequesters Nrf2 in the cytoplasm under un-stimulated conditions, were unchanged. Translocation of Nrf2 into the nuclei of hepatocytes was observed using western blotting and immunohistochemistry. Expression of mRNAs for Nrf2-dependent antioxidant and phase II enzymes, such as heme oxygenase 1 (HO-1), NAD (P) H dehydrogenase, quinone 1 (Nqo1), glutathione S-transferase A2 (Gsta2), UDP glucuronosyltransferase 1 family polypeptide A6 (Ugt1a6), were dose-dependently up-regulated at 3 h. Furthermore, the levels of HO-1 IR were dose-dependently increased in hepatocytes at 6 h. Subcutaneous administration of lansoprazole (30 mg/kg/day) for 7 successive days resulted in up-regulation and nuclear translocation of Nrf2 IR in hepatocytes and up-regulation of HO-1 IR in the liver. Pretreatment with lansoprazole attenuated thioacetamide (500 mg/kg)-induced acute hepatic damage via both HO-1-dependent and -independent pathways. Up-stream networks related to Nrf2 expression were investigated using microarray analysis, followed by data mining with Ingenuity Pathway Analysis. Up-regulation of the aryl hydrocarbon receptor (AhR)-cytochrome P450, family 1, subfamily a, polypeptide 1 (Cyp1a1) pathway was associated with up-regulation of Nrf2 mRNA. In conclusion, lansoprazole might have an alternative indication in the prevention and treatment of oxidative hepatic damage through the induction of both phase I and phase II drug-metabolizing systems, i.e. the AhR/Cyp1a1/Nrf2 pathway in hepatocytes.
Sigma-Aldrich
Lansoprazole, ≥98% (TLC), powder
Lansoprazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Lansoprazole, European Pharmacopoeia (EP) Reference Standard
USP
Lansoprazole, United States Pharmacopeia (USP) Reference Standard