Merck
  • Abnormal splicing of NEDD4 in myotonic dystrophy type 2: possible link to statin adverse reactions.
Abnormal splicing of NEDD4 in myotonic dystrophy type 2: possible link to statin adverse reactions.
The American journal of pathology (2014-06-08)
Mark Screen, Per Harald Jonson, Olayinka Raheem, Johanna Palmio, Reijo Laaksonen, Terho Lehtimäki, Mario Sirito, Ralf Krahe, Peter Hackman, Bjarne Udd
ABSTRACT
Myotonic dystrophy type 2 (DM2) is a multisystemic disorder caused by a (CCTG)n repeat expansion in intron 1 of CNBP. Transcription of the repeats causes a toxic RNA gain of function involving their accumulation in ribonuclear foci. This leads to sequestration of splicing factors and alters pre-mRNA splicing in a range of downstream effector genes, which is thought to contribute to the diverse DM2 clinical features. Hyperlipidemia is frequent in DM2 patients, but the treatment is problematic because of an increased risk of statin-induced adverse reactions. Hypothesizing that shared pathways lead to the increased risk, we compared the skeletal muscle expression profiles of DM2 patients and controls with patients with hyperlipidemia on statin therapy. Neural precursor cell expressed, developmentally downregulated-4 (NEDD4), an ubiquitin ligase, was one of the dysregulated genes identified in DM2 patients and patients with statin-treated hyperlipidemia. In DM2 muscle, NEDD4 mRNA was abnormally spliced, leading to aberrant NEDD4 proteins. NEDD4 was down-regulated in persons taking statins, and simvastatin treatment of C2C12 cells suppressed NEDD4 transcription. Phosphatase and tensin homologue (PTEN), an established NEDD4 target, was increased and accumulated in highly atrophic DM2 muscle fibers. PTEN ubiquitination was reduced in DM2 myofibers, suggesting that the NEDD4-PTEN pathway is dysregulated in DM2 skeletal muscle. Thus, this pathway may contribute to the increased risk of statin-adverse reactions in patients with DM2.
Sigma-Aldrich
Anti-PTEN Antibody, clone 6H2.1, clone 6H2.1, Upstate®, from mouse
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Simvastatin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Supelco
Simvastatin, analytical standard