Merck
  • Are growth factor receptors modulated by metformin in human endometrial stromal cells after stimulation with androgen and insulin?
Are growth factor receptors modulated by metformin in human endometrial stromal cells after stimulation with androgen and insulin?
Archives of gynecology and obstetrics (2014-03-15)
Gustavo Dias Ferreira, Ariane Germeyer, Amanda de Barros Machado, Tadeu Ludwig do Nascimento, Ilma Simoni Brum, Thomas Strowitzki, Helena von Eye Corleta, Edison Capp
ABSTRACT
To assess the effect of metformin on gene and protein expression of insulin receptor (IR) and IGF-1 (IGF-1R) receptor in human endometrial stromal cells after stimulation with androgen and insulin. Primary culture of endometrial stromal cells stimulated with estrogen, progesterone with or without androgen or insulin, and treated with metformin for 24 and 48 h, followed by RNA (qRT-PCR) and protein (Western blot) extraction and analysis. IR gene expression was increased after treatment with insulin (2.9-fold change, p = 0.027) and further after metformin treatment (4.7-fold change, p < 0.001), and in IGF-1R, the group treated with insulin (1.83-fold change) and metformin (1.78-fold change) showed more expression, than control group (p < 0.001). Similarly, IR protein expression was increased after addition of metformin and insulin (249,869 ± 15,878) in relation to the other groups (p < 0.001). Furthermore, cells treated with insulin (153,634 ± 29,123) and androgen plus insulin (162,854 ± 86,258) had a higher IR protein expression compared to control (104,654 ± 5,634) and androgen group (71,595 ± 3,439, (p = 0.045 and 0.021). In groups treated with insulin (127,711 ± 4,591) and androgen plus insulin (151,098 ± 5,194) the protein IGF-1R was increased compared to control (79,355 ± 3,470) and the androgen-only group (79,326 ± 3,114) (p < 0.001). Metformin in combination with insulin increased IR protein and gene expressions, while it had no influence on the protein expression of IGF-1R in endometrial stromal cells.
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, natural, 97%, FG
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material