Merck
  • Genotoxic assessment of Rubus imperialis (Rosaceae) extract in vivo and its potential chemoprevention against cyclophosphamide-induced DNA damage.
Genotoxic assessment of Rubus imperialis (Rosaceae) extract in vivo and its potential chemoprevention against cyclophosphamide-induced DNA damage.
Journal of ethnopharmacology (2014-04-02)
Ana Beatriz Costa Rodrigues Alves, Rafaella Souza dos Santos, Susana de Santana Calil, Rivaldo Niero, Jhonny da Silva Lopes, Fábio F Perazzo, Paulo César Pires Rosa, Sérgio Faloni Andrade, Valdir Cechinel-Filho, Edson Luis Maistro
ABSTRACT
Rubus imperialis Cham. Schl. (Rosaceae) is frequently used in traditional medicine as hypoglycemic, antinociceptive and antiviral remedy. Swiss albino mice were distributed in eight groups for acute treatment with Rubus imperialis extract (24 h). The extract doses selected were 50, 250 and 500 mg/kg b.w. administered by gavage alone or plus to CPA (50 mg/kg b.w.) administered by intraperitoneal injection. Control groups were treated in a similar way. Analyses were performed using the comet assay, on leukocytes (collected 4 and 24h after treatment) and liver (collected 24 h after treatment), and using the micronucleus test (MN) in bone marrow cells. Cytotoxicity was assessed by scoring 200 consecutive polychromatic (PCE) and normochromatic (NCE) erythrocytes (PCE/NCE ratio). The main compounds identified in the Rubus imperialis extract were saponins and steroidal compounds, with niga-ichigoside and tormentic acid being the major compounds. Tested doses of Rubus imperialis extract showed no genotoxic effects on leukocytes from peripheral blood or liver cells by the comet assay. However, the MN test showed an increase in the frequency of micronucleated cells at the two higher doses tested, indicating that this extract has clastogenic/aneugenic effects on bone marrow cells at higher doses. On the other hand, for all cells evaluated, the three tested doses of the Rubus imperialis extract promoted inhibition of DNA damage induced by CPA. Despite the chemoprevention observed, the clastogenicity/aneugenicity observed suggested caution about either continuous or high-dose usage of Rubus imperialis aerial parts extract by humans.
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, BioUltra, for molecular biology, ≥99.0% (HPLC)
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, detergent for use in cell lysis
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, ≥94%
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, BioXtra, ≥97% (TLC)
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, ≥97.0% (HPLC)
Sigma-Aldrich
N-Lauroylsarcosine sodium salt solution, 20%, for molecular biology
Sigma-Aldrich
N-Lauroylsarcosine sodium salt solution, 30% aqueous solution, ≥97.0% (HPLC)
Cyclophosphamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder