Merck
  • An analysis of 72% chromated glycerin used for sclerotherapy: sterility, potency, and cost after extended shelf life.
An analysis of 72% chromated glycerin used for sclerotherapy: sterility, potency, and cost after extended shelf life.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.] (2014-12-19)
Amir M Ghaznavi, Mio Nakamura, Donna Tepper
ABSTRACT
Sclerotherapy is the treatment of reticular veins and telangiectasias of the lower extremities. Sclerosants destroy endothelial tissue and expose subendothelial collagen fibers, which lead to subsequent fibrosis of vessels, thus preventing recanalization. There are several available sclerosants including sodium tetradecyl sulfate (STS), polidocanol (POL), and chromated glycerin (CG) with varying efficacy, potency, side effect profile, and cost. To identify the possible bacterial contamination and potency of CG beyond the current recommended shelf life of 3 months and to prove if CG is as cost effective as other available sclerosants. Samples of 72% CG underwent bacterial endotoxin, sterility, and potency analysis at Days 0, 24, and 183. In addition, cost comparison was performed with other commercially available sclerosants including STS and POL. No samples of CG showed any bacterial contamination. All aliquots of glycerin remained sterile at Day 14. Potency at Day 24 was 99.2%, which was the same at Day 183. Cost comparison with other sclerosants revealed that CG is lower cost per milliliter than STS and POL. Seventy-two percent CG has no contamination and maintains its reported potency up to 6 months while comparable with the cost of other commercially available sclerosants.
Sigma-Aldrich
Tetradecyl sulfate sodium salt, 95%
Sigma-Aldrich
Thesit®, for membrane research
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Brij® L23 solution, 30 % (w/v) in H2O
Sigma-Aldrich
Decaethylene glycol mono­dodecyl ether, nonionic surfactant
Sigma-Aldrich
ECO Brij® L23
Sigma-Aldrich
ECO BRIJ® L4, average Mn ~362
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Brij® L4, average Mn ~362
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Brij® L23, suitable for Stein-Moore chromatography
Sigma-Aldrich
Brij® L23, main component: tricosaethylene glycol dodecyl ether
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)