Merck
  • Comprehensive therapy for hemangioma presenting with Kasabach-Merritt syndrome in the maxillofacial region.
Comprehensive therapy for hemangioma presenting with Kasabach-Merritt syndrome in the maxillofacial region.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2014-12-17)
Lixin Su, Deming Wang, Xindong Fan
ABSTRACT
To summarize the characteristics of Kasabach-Merritt syndrome (KMS) and to evaluate the therapeutic effect of drug therapy combined with transarterial embolization. From 2007 to 2011, 6 cases of KMS that underwent drug therapy and transarterial embolization were retrospectively analyzed, comprising of 3 male and 3 female patients; the ages of the patients ranged from 3 to 40 days. The lesions were located in the temporal region (1 of 6, 16.7%), parotid region (2 of 6, 33.3%), or submandibular region and neck (3 of 6, 50%). All the patients were followed for 12 to 18 months. Therapeutic outcomes were assessed by evaluating the platelet count, coagulation parameters, and size of the lesion. Positive responses were visible shrinkage of the hemangioma or lightening of the skin color of the cutaneous tumor (or both) within 8 to 72 hours in 4 patients (66.67%). These occurred within 1 week in 5 patients (83.33%) and within 2 weeks in all patients (100%). The mean platelet count before treatment was 18,000/L (range, 8,000 to 33,000/L). After the first week of medical treatment, the mean platelet count increased to above 80,000/L in 5 patients (83.33%); it began to increase on the second day after embolization and reached 102,000/L in 1 patient (16.67%). All 6 cases (100%) showed good results with treatment. A 12- to 18-month follow-up evaluation was obtained for all treated patients, and no rebound growth or platelet count decreases were observed. No severe or obvious adverse complications were noted during all treatment courses. For most hemangiomas presenting with KMS, good results can be obtained with systemic medical treatment. Transcatheter arterial embolization with polyvinyl alcohol particles combined with systemic medical treatment should be considered an efficacious and important therapeutic option for challenging cases.
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 8,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
USP
Propranolol hydrochloride, United States Pharmacopeia (USP) Reference Standard
Poly(vinyl alcohol), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
6α-Methylprednisolone, ≥98%
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Supelco
(±)-Propranolol hydrochloride, analytical standard