Merck
  • Characterization of cadmium plasma membrane transport in gills of a mangrove crab Ucides cordatus.
Characterization of cadmium plasma membrane transport in gills of a mangrove crab Ucides cordatus.
Aquatic toxicology (Amsterdam, Netherlands) (2014-12-03)
P Ortega, M R Custódio, F P Zanotto
ABSTRACT
Membrane pathway for intracellular cadmium (Cd(2+)) accumulation is not fully elucidated in many organisms and has not been studied in crab gill cells. To characterize membrane Cd(2+) transport of anterior and posterior gill cells of Ucides cordatus, a hypo-hyper-regulating crab, a change in intracellular Cd(2+) concentration under various experimental conditions was examined by using FluoZin, a fluorescent probe. The membrane Cd(2+) transport was estimated by the augmentation of FluoZin fluorescence induced by extracellular application of CdCl2 and different inhibitors. Addition of extracellular calcium (Ca(2+)) to the cells affected little the fluorescence of FluoZin, confirming that Cd(2+) was the main ion increasing intracellular fluorescence. Ca(2+) channels blockers (nimodipine and verapamil) decreased Cd(2+) influx as well as vanadate, a Ca(2+)-ATPase blocker. Chelating intracellular Ca(2+) (BAPTA) decreased Cd(2+) influx in gill cells, while increasing intracellular Ca(2+) (caffeine) augmented Cd influx. Cd(2+) and ATP added at different temporal conditions were not effective at increasing intracellular Cd(2+) accumulation. Ouabain (Na(+)/K(+)-ATPase inhibitor) increased Cd(2+) influx probably through a change in intracellular Na and/or a change in cell membrane potential. Routes of Cd(2+) influx, a non-essential metal, through the gill cell plasma membrane of crabs are suggested.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Caffeine, European Pharmacopoeia (EP) Reference Standard
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
1,2-Bis(2-Aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid, 98%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous