Merck
  • The effect of phthalate exposure and filaggrin gene variants on atopic dermatitis.
The effect of phthalate exposure and filaggrin gene variants on atopic dermatitis.
Environmental research (2014-12-03)
I-Jen Wang, Wilfried Joachim Jurgen Karmaus
ABSTRACT
Phthalate exposure may increase the risk of atopic disorders. However, little is known about the joint effects of phthalate exposure and filaggrin (FLG) gene variants on atopic dermatitis (AD). We want to investigate whether FLG variants are related to a higher urine concentration of phthalates and whether an interaction of FLG and phthalates increases the risk of AD. We conducted a case-control study comprised of 106 AD children and 347 controls, all of whom were selected from CEAS cohort. Urine phthalate metabolite levels (MEP, MBP, MBzP, and 5OH-MEHP) were measured by UPLC-MS/MS. FLG variants were analyzed by TaqMan assay. At 3 years of age, information about the development of AD and environmental exposures were collected. Logistic regressions were performed to estimate the association of genotypes and phthalate metabolite levels with AD. Urine MBP and MBzP levels were higher in children with AD than in controls (p<0.001). Children with the FLG P478S TT genotype had higher urine phthalate metabolite levels as compared with CC carriers, with MBP and MBzP having a statistically significant difference (geometric mean(s.e.) 5.51(3.77) vs. 3.03(3.48), p=0.015 and 0.76(3.01) vs. 0.53(2.56), p=0.018). After stratifying by phthalate metabolite levels, FLG P478S TT genotype was related to a higher odds of AD in children with high MBP levels (aOR=4.74, 95% CI 1.45-15.5) and in children with high MBzP levels (aOR=3.46, 95%CI 1.03-11.58). FLG variants may increase skin permeability leading to higher skin absorption of phthalate and thus confer a higher susceptibility for AD. Or alternatively, the internal burden of phthalates metabolites is increased in children with AD who also have risky variant of the FLG gene.
Product Description
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Phthalic acid dipotassium salt, 98%
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
mono-Butyl phthalate, 97.0-103.0% (T)
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Supelco
mono-Butyl phthalate, analytical standard
Phthalic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Phthalic acid, analytical standard
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)