Merck
  • The contribution of activating transcription factor 3 to apoptosis of human colorectal cancer cells by protocatechualdehyde, a naturally occurring phenolic compound.
The contribution of activating transcription factor 3 to apoptosis of human colorectal cancer cells by protocatechualdehyde, a naturally occurring phenolic compound.
Archives of biochemistry and biophysics (2014-12-03)
Jeong Rak Lee, Man Hyo Lee, Hyun Ji Eo, Gwang Hun Park, Hun Min Song, Mi Kyoung Kim, Jin Wook Lee, Jin Boo Jeong
ABSTRACT
Protocatechualdehyde (PCA) is one of the important compounds found in barley, green cavendish bananas and grapevine leaves. PCA shows anti-cancer activities in breast, leukemia and colorectal cancer cells. Previous study reported that PCA exerts anti-cancer activity through down-regulating cyclin D1 and HDAC2 in human colorectal cancer cells. However, the underlying mechanisms for the expression of activating transcription factor 3 (ATF3) by PCA has not been studied. Thus, we performed in vitro study to investigate if treatment of PCA affects ATF3 expression and ATF3-mediated apoptosis in human colorectal cancer cells. PCA decreased cell viability in a dose-dependent manner in HCT116 and SW480 cells. In addition, PCA reduced cell viability in MCF-7, MDA-MB-231 and HepG-2 cells. Exposure of PCA activated the levels of ATF3 protein and mRNA in HCT116 and SW480 cells. Inhibition of ERK1/2/ by PD98059 and p38 by SB203580 inhibited PCA-induced ATF3 expression and transcriptional activation. ATF3-knockdown inhibited PCA-induced apoptosis and cell viability. In addition, ATF3 overexpression enhanced PCA-mediated cleavage of PARP. These findings suggest that inhibition of cell viability and apoptosis by PCA may be result of ATF3 expression through ERK1/2 and p38-mediated transcriptional activation.
Sigma-Aldrich
3,4-Dihydroxybenzaldehyde, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
3,4-Dihydroxybenzaldehyde, 97%
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%
Supelco
3,4-Dihydroxybenzaldehyde, analytical standard
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, packed in coated, shock- and leak-protected glass bottle, ≥60% (T)
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%