Merck
  • Spectroscopic studies of the Salmonella enterica adenosyltransferase enzyme SeCobA: molecular-level insight into the mechanism of substrate Cob(II)alamin activation.
Spectroscopic studies of the Salmonella enterica adenosyltransferase enzyme SeCobA: molecular-level insight into the mechanism of substrate Cob(II)alamin activation.
Biochemistry (2014-11-26)
Ivan G Pallares, Theodore C Moore, Jorge C Escalante-Semerena, Thomas C Brunold
ABSTRACT
CobA from Salmonella enterica (SeCobA) is a member of the family of ATP:Co(I)rrinoid adenosyltransferase (ACAT) enzymes that participate in the biosynthesis of adenosylcobalamin by catalyzing the transfer of the adenosyl group from an ATP molecule to a reactive Co(I)rrinoid species transiently generated in the enzyme active site. This reaction is thermodynamically challenging, as the reduction potential of the Co(II)rrinoid precursor in solution is far more negative than that of available reducing agents in the cell (e.g., flavodoxin), precluding nonenzymic reduction to the Co(I) oxidation state. However, in the active sites of ACATs, the Co(II)/Co(I) redox potential is increased by >250 mV via the formation of a unique four-coordinate (4c) Co(II)rrinoid species. In the case of the SeCobA ACAT, crystallographic and kinetic studies have revealed that the phenylalanine 91 (F91) and tryptophan 93 (W93) residues are critical for in vivo activity, presumably by blocking access to the lower axial ligand site of the Co(II)rrinoid substrate. To further assess the importance of the F91 and W93 residues with respect to enzymatic function, we have characterized various SeCobA active-site variants using electronic absorption, magnetic circular dichroism, and electron paramagnetic resonance spectroscopies. Our data provide unprecedented insight into the mechanism by which SeCobA converts the Co(II)rrinoid substrate to 4c species, with the hydrophobicity, size, and ability to participate in offset π-stacking interactions of key active-site residues all being critical for activity. The structural changes that occur upon Co(II)rrinoid binding also appear to be crucial for properly orienting the transiently generated Co(I) "supernucleophile" for rapid reaction with cosubstrate ATP.
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Cobalt, foil, 10mm disks, thickness 0.0125mm, 99.9%
Cobalt, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Cobalt, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Cobalt, foil, 15mm disks, thickness 0.02mm, 99.9%
Cobalt, foil, 250x250mm, thickness 0.50mm, annealed, 99.9%
Cobalt, wire reel, 0.5m, diameter 0.125mm, hard, 99.99+%
Cobalt, foil, 15mm disks, thickness 0.0125mm, 99.9%
Cobalt, foil, 10mm disks, thickness 0.015mm, 99.9%
Cobalt, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cobalt, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%
Cobalt, foil, 50x50mm, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, 10mm disks, thickness 0.005mm, 99.9%
Cobalt, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Cobalt, foil, 10mm disks, thickness 0.01mm, 99.9%
Cobalt, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Cobalt, wire reel, 0.1m, diameter 0.05mm, hard, 99.9%
Cobalt, foil, 10mm disks, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%