Merck
  • Splicing factor SR34b mutation reduces cadmium tolerance in Arabidopsis by regulating iron-regulated transporter 1 gene.
Splicing factor SR34b mutation reduces cadmium tolerance in Arabidopsis by regulating iron-regulated transporter 1 gene.
Biochemical and biophysical research communications (2014-12-03)
Wentao Zhang, Bojing Du, Di Liu, Xiaoting Qi
ABSTRACT
Serine/arginine-rich (SR) proteins are important splicing factors. However, the biological functions of plant SR proteins remain unclear especially in abiotic stresses. Cadmium (Cd) is a non-essential element that negatively affects plant growth and development. In this study, we provided clear evidence for SR gene involved in Cd tolerance in planta. Systemic expression analysis of 17 Arabidopsis SR genes revealed that SR34b is the only SR gene upregulated by Cd, suggesting its potential roles in Arabidopsis Cd tolerance. Consistent with this, a SR34b T-DNA insertion mutant (sr34b) was moderately sensitive to Cd, which had higher Cd(2+) uptake rate and accumulated Cd in greater amounts than wild-type. This was due to the altered expression of iron-regulated transporter 1 (IRT1) gene in sr34b mutant. Under normal growth conditions, IRT1 mRNAs highly accumulated in sr34b mutant, which was a result of increased stability of IRT1 mRNA. Under Cd stress, however, sr34b mutant plants had a splicing defect in IRT1 gene, thus reducing the IRT1 mRNA accumulation. Despite of this, sr34b mutant plants still constitutively expressed IRT1 proteins under Cd stress, thereby resulting in Cd stress-sensitive phenotype. We therefore propose the essential roles of SR34b in posttranscriptional regulation of IRT1 expression and identify it as a regulator of Arabidopsis Cd tolerance.
Product Description
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Iron, rod, 1000mm, diameter 30mm, as drawn, 98+%
Iron, rod, 150mm, diameter 5.0mm, as drawn, 99.99+%
Iron, foil, 100x100mm, thickness 0.15mm, hard, 99.5%
Iron, rod, 200mm, diameter 8.0mm, as drawn, 99.99+%
Iron, foil, light tested, 50x50mm, thickness 0.025mm, hard, 99.5%
Iron, foil, light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Iron, rod, 100mm, diameter 4.8mm, as drawn, 98+%
Iron, foil, not light tested, 50x50mm, thickness 0.002mm, 99.85%
Iron, rod, 100mm, diameter 100mm, as drawn, armcO« soft ingot 99.8%
Iron, rod, 500mm, diameter 19mm, as drawn, soft ingot 99.8%
Iron, foil, light tested, 25x25mm, thickness 0.075mm, hard, 99.5%
Iron, rod, 1000mm, diameter 12.7mm, as drawn, 98+%
Iron, foil, 50mm disks, thickness 0.38mm, hard, 99.5%
Iron, rod, 500mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%
Iron, foil, not light tested, 150x150mm, thickness 0.009mm, 99.85%
Iron, rod, 100mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%
Iron, foil, not light tested, 50x50mm, thickness 0.01mm, 99.99+%
Iron, rod, 1000mm, diameter 19.0mm, as drawn, 98+%
Iron, foil, 150x150mm, thickness 0.38mm, hard, 99.5%