Merck
  • Mechanism and treatment for learning and memory deficits in mouse models of Noonan syndrome.
Mechanism and treatment for learning and memory deficits in mouse models of Noonan syndrome.
Nature neuroscience (2014-11-11)
Yong-Seok Lee, Dan Ehninger, Miou Zhou, Jun-Young Oh, Minkyung Kang, Chuljung Kwak, Hyun-Hee Ryu, Delana Butz, Toshiyuki Araki, Ying Cai, J Balaji, Yoshitake Sano, Christine I Nam, Hyong Kyu Kim, Bong-Kiun Kaang, Corinna Burger, Benjamin G Neel, Alcino J Silva
ABSTRACT
In Noonan syndrome (NS) 30-50% of subjects show cognitive deficits of unknown etiology and with no known treatment. Here, we report that knock-in mice expressing either of two NS-associated mutations in Ptpn11, which encodes the nonreceptor protein tyrosine phosphatase Shp2, show hippocampal-dependent impairments in spatial learning and deficits in hippocampal long-term potentiation (LTP). In addition, viral overexpression of an NS-associated allele PTPN11(D61G) in adult mouse hippocampus results in increased baseline excitatory synaptic function and deficits in LTP and spatial learning, which can be reversed by a mitogen-activated protein kinase kinase (MEK) inhibitor. Furthermore, brief treatment with lovastatin reduces activation of the GTPase Ras-extracellular signal-related kinase (Erk) pathway in the brain and normalizes deficits in LTP and learning in adult Ptpn11(D61G/+) mice. Our results demonstrate that increased basal Erk activity and corresponding baseline increases in excitatory synaptic function are responsible for the LTP impairments and, consequently, the learning deficits in mouse models of NS. These data also suggest that lovastatin or MEK inhibitors may be useful for treating the cognitive deficits in NS.
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Lovastatin, European Pharmacopoeia (EP) Reference Standard
USP
Lovastatin, United States Pharmacopeia (USP) Reference Standard
Supelco
Lovastatin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Lovastatin for peak identification, European Pharmacopoeia (EP) Reference Standard