Merck
  • Structures of bacterial polynucleotide kinase in a michaelis complex with nucleoside triphosphate (NTP)-Mg2+ and 5'-OH RNA and a mixed substrate-product complex with NTP-Mg2+ and a 5'-phosphorylated oligonucleotide.
Structures of bacterial polynucleotide kinase in a michaelis complex with nucleoside triphosphate (NTP)-Mg2+ and 5'-OH RNA and a mixed substrate-product complex with NTP-Mg2+ and a 5'-phosphorylated oligonucleotide.
Journal of bacteriology (2014-10-01)
Ushati Das, Li Kai Wang, Paul Smith, Annum Munir, Stewart Shuman
ABSTRACT
Clostridium thermocellum polynucleotide kinase (CthPnk), the 5'-end-healing module of a bacterial RNA repair system, catalyzes reversible phosphoryl transfer from a nucleoside triphosphate (NTP) donor to a 5'-OH polynucleotide acceptor, either DNA or RNA. Here we report the 1.5-Å crystal structure of CthPnk-D38N in a Michaelis complex with GTP-Mg(2+) and a 5'-OH RNA oligonucleotide. The RNA-binding mode of CthPnk is different from that of the metazoan RNA kinase Clp1. CthPnk makes hydrogen bonds to the ribose 2'-hydroxyls of the 5' terminal nucleoside, via Gln51, and the penultimate nucleoside, via Gln83. The 5'-terminal nucleobase is sandwiched by Gln51 and Val129. Mutating Gln51 or Val129 to alanine reduced kinase specific activity 3-fold. Ser37 and Thr80 donate functionally redundant hydrogen bonds to the terminal phosphodiester; a S37A-T80A double mutation reduced kinase activity 50-fold. Crystallization of catalytically active CthPnk with GTP-Mg(2+) and a 5'-OH DNA yielded a mixed substrate-product complex with GTP-Mg(2+) and 5'-PO4 DNA, wherein the product 5' phosphate group is displaced by the NTP γ phosphate and the local architecture of the acceptor site is perturbed.
Sigma-Aldrich
Polynucleotide Kinase from T4-infected Escherichia coli, 10 units/μL, buffered aqueous glycerol solution
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, microfoil, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Magnesium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, microfoil, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, microleaf, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Magnesium, wire reel, 1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Magnesium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.25mm, as drawn, 99.9+%