Merck
  • Oxidative stability of n-3-enriched chicken patties under different package-atmosphere conditions.
Oxidative stability of n-3-enriched chicken patties under different package-atmosphere conditions.
Food chemistry (2014-08-31)
Ana Penko, Tomaž Polak, Mateja Lušnic Polak, Tomaž Požrl, Damir Kakovič, Božidar Žlender, Lea Demšar
ABSTRACT
The oxidation processes were studied in chicken patties, enriched with n-3 fatty acids, after 8days of storage at 4°C, under different aerobic conditions, and following heat treatment. Significant effects were seen on lipid and cholesterol oxidation and the sensory qualities for whole flaxseed addition in the chicken feed (i.e., n-3 fatty acid enrichment), and for the different package-atmosphere conditions. For the raw chicken patties, n-3 enrichment increased the colour L(∗) values while, after the heat treatment, there were higher thiobarbituric acid-reactive substances (TBARs) and cholesterol oxidation products (COPs), and the rancidity was more pronounced. In comparison with the low O2 (<0.5%) package-atmosphere condition, O2 enrichment (80%) increased the instrumentally measured colour values, TBARs, total and individual COPs, and the rancidity became pronounced. The most suitable package-atmosphere condition of these raw n-3-enriched chicken patties is a very low O2 atmosphere, with or without an O2 scavenger.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
25-Hydroxycholesterol, ≥98%
Sigma-Aldrich
Methyl linoleate, ≥98% (GC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methyl linoleate, analytical standard
USP
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard