Merck
  • Doublecortin-expressing cells are present in layer II across the adult guinea pig cerebral cortex: partial colocalization with mature interneuron markers.
Doublecortin-expressing cells are present in layer II across the adult guinea pig cerebral cortex: partial colocalization with mature interneuron markers.
Experimental neurology (2008-04-02)
Kun Xiong, Duan-Wu Luo, Peter R Patrylo, Xue-Gang Luo, Robert G Struble, Richard W Clough, Xiao-Xin Yan
ABSTRACT
Doublecortin-immunoreactive (DCX+) cells were detected across the allo- and neo-cortical regions in the adult guinea pig cerebrum, localized to layer II specifically at its border with layer I. The density of labeled cells declined with age, whereas no apparent apoptotic activity was detectable over the cortex including layer II. DCX+ cells varied in somal size, labeling intensity, nuclear appearance, and complexity of processes. These cells were often arranged in clusters with cells of similar morphology sometimes packed tightly together. They exhibited complete colocalization with polysialylated neural cell adhesion molecule (PSA-NCAM) and neuron-specific type III beta-tubulin (TuJ1). Medium to large-sized DCX+ cells had well-developed neuritic processes, and expressed neuron-specific nuclear protein (NeuN). Large mature-looking cells with weak DCX reactivity invariably displayed heavy NeuN reactivity, implicating a transitional stage of these labeled cells. These "transitional" cells also consistently exhibited weak reactivity for gamma-aminobutyric acid (GABA), glutamate decarboxylase (GAD67), beta-nicotinamide adenine dinucleotide phosphate diaphorase (NADPH-d) and neuronal nitric oxide synthase (nNOS), suggestive of them being young GABAergic/nitrinergic interneurons. Our data indicate that DCX+ cells exist widely in the adult guinea pig cerebral cortex, with a predominant localization in upper layer II. The morphological variation and differential expression of neuronal markers in these cells implicate that they might be developing neurons, and that they are probably differentiating into GABAergic interneurons. This population of cells might be involved in interneuron plasticity in the adult mammalian cerebral cortex.
Sigma-Aldrich
Monoclonal Anti-Parvalbumin antibody produced in mouse, clone PARV-19, ascites fluid
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Polysialic Acid-NCAM Antibody, clone 2-2B, ascites fluid, clone 2-2B, Chemicon®
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
Anti-Nitric Oxide Synthase I Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate, ≥97% (HPLC)
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-189 mreelin, clone 142, clone 142, Chemicon®, from mouse