Merck
  • Femtosecond study of self-trapped vibrational excitons in crystalline acetanilide.
Femtosecond study of self-trapped vibrational excitons in crystalline acetanilide.
Physical review letters (2002-02-28)
J Edler, P Hamm, A C Scott
ABSTRACT
Femtosecond IR spectroscopy of delocalized NH excitations of crystalline acetanilide confirms that self-trapping in hydrogen-bonded peptide units exists and does stabilize the excitation. Two phonons with frequencies of 48 and 76 cm (-1) are identified as the major degrees of freedom that mediate self-trapping. After selective excitation of the free exciton, self-trapping occurs within a few 100 fs. Excitation of the self-trapped states disappears from the spectral window of this investigation on a 1 ps time scale, followed by a slow ground state recovery of the hot ground state within 18 ps.
Sigma-Aldrich
Acetanilide, zone-refined, purified by sublimation, ≥99.95%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetanilide, purified by sublimation, ≥99.9%
Pricing and availability is not currently available.
USP
Acetanilide Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.