- High-performance liquid chromatographic separation and on-line mass spectrometric detection of saturated and unsaturated oligogalacturonic acids.
High-performance liquid chromatographic separation and on-line mass spectrometric detection of saturated and unsaturated oligogalacturonic acids.
Carbohydrate research (2002-12-21)
Thomas Stoll, Andreas Schieber, Reinhold Carle
PMID12493233
ABSTRACT
A method for the simultaneous determination of saturated and unsaturated oligogalacturonic acids up to degree of polymerization (dp) of 7 by high-performance liquid chromatography (HPLC) is presented. For this purpose, a Cyclobond I 2000 column and a volatile mobile phase consisting of ammonium formate and methanol were used, allowing direct coupling of HPLC to a mass spectrometer via an electrospray interface (ESI-MS) without additional desalting. The analytical system was used for the characterization of digests obtained by incubation of polygalacturonic acid with commercial enzyme preparations.
Product Description
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm