- Synthesis of chiral 3-alkyl-3,4-dihydroisocoumarins by dynamic kinetic resolutions catalyzed by a Baeyer-Villiger monooxygenase.
Synthesis of chiral 3-alkyl-3,4-dihydroisocoumarins by dynamic kinetic resolutions catalyzed by a Baeyer-Villiger monooxygenase.
The Journal of organic chemistry (2010-02-20)
Ana Rioz-Martínez, Gonzalo de Gonzalo, Daniel E Torres Pazmiño, Marco W Fraaije, Vicente Gotor
PMID20166716
ABSTRACT
Baeyer-Villiger monooxygenases have been tested in the oxidation of racemic benzofused ketones. When employing a single mutant of phenylacetone monooxygenase (M446G PAMO) under the proper reaction conditions, it was possible to achieve 3-substituted 3,4-dihydroisocoumarins with high yields and optical purities through regioselective dynamic kinetic resolution processes.