Merck
  • Dodecaamide cages: organic 12-arm building blocks for supramolecular chemistry.
Dodecaamide cages: organic 12-arm building blocks for supramolecular chemistry.
Journal of the American Chemical Society (2013-06-22)
Jamie L Culshaw, Ge Cheng, Marc Schmidtmann, Tom Hasell, Ming Liu, Dave J Adams, Andrew I Cooper
ABSTRACT
A simple, one-step amidation reaction is used to produce a range of 12-arm organic building blocks for supramolecular chemistry via the derivatization of porous imine cages. As an example, microporous dendrimers are prepared.