Merck
  • Selective synthesis of α,α-dideuterio alcohols by the reduction of carboxylic acids using SmI2 and D2O as deuterium source under SET conditions.
Selective synthesis of α,α-dideuterio alcohols by the reduction of carboxylic acids using SmI2 and D2O as deuterium source under SET conditions.
Organic letters (2014-09-24)
Michal Szostak, Malcolm Spain, David J Procter
ABSTRACT
The first general method for the chemoselective synthesis of α,α-dideuterio alcohols directly from feedstock carboxylic acids under single electron transfer conditions using SmI2 is reported. This reaction proceeds after the activation of Sm(II) with a Lewis base, results in excellent levels of deuterium incorporation across a wide range of substrates, and represents an attractive alternative to processes mediated by pyrophoric alkali metal deuterides.
Sigma-Aldrich
Samarium, −40 mesh, 99% trace rare earth metals basis
Samarium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Samarium, rod, 100mm, diameter 6.35mm, cast, 99%
Samarium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Samarium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Samarium, rod, 50mm, diameter 6.35mm, cast, 99%