Merck
  • Bioavailability of cadmium and biochemical responses on the freshwater bivalve Corbicula fluminea--the role of TiO₂ nanoparticles.
Bioavailability of cadmium and biochemical responses on the freshwater bivalve Corbicula fluminea--the role of TiO₂ nanoparticles.
Ecotoxicology and environmental safety (2014-09-10)
Gonçalo Vale, Cristiana Franco, Mário S Diniz, Margarida M C dos Santos, Rute F Domingos
ABSTRACT
The increasing and widespread applications of TiO2 engineered nanoparticles (nTiO2) led to the release of these materials into aquatic environments and consequently a change on the assessment of the environmental risk of trace metals. In this work, the role of two commercial nTiO2 with distinct crystalline phases and sizes (nTiO2-P25: 80% anatase+20% rutile, d=20nm; nTiO2-NA: 100% anatase, d=5 nm; 0.1 and 1.0 mg L(-1)) on Cd (112 μg L(-1)) speciation, biouptake and toxicity for the freshwater bivalve Corbicula fluminea was evaluated. The electroanalytical technique 'absence of gradients and Nernstian equilibrium stripping (AGNES)' was used to quantify the free Cd concentrations in the exposure medium in presence of both particles. Despite ca. 30-40% decrease of free Cd in the medium in presence of nTiO2, Cd uptake by C. fluminea was similar in the absence and presence of either of the particles. Superoxide dismutase and glutathione-S-transferase activities remained unchanged for Cd in absence and presence of nTiO2, whereas a significant increase of the catalase activity was obtained at the third day for Cd in presence of both nTiO2. Despite lipid peroxidation data shows that the presence of both nTiO2 seems to exert cells damage, a more quantitative description is not possible with the obtained data. The lack of clear-cut responses by the studied biomarkers, even when only in presence of Cd, do not allow insights into the effect of the presence of nTiO2 on the Cd toxicity to the bivalves. Notwithstanding, morphological changes in the digestive gland were clearly obtained in the presence of Cd, nTiO2 and Cd+nTiO2 indicating an inflammatory response.
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, sponge, 1-20 mm, 99.5% trace metals basis
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Titanium, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Glutathione S-Transferase from equine liver, lyophilized powder, ≥25 units/mg protein
Cadmium, foil, 6mm disks, thickness 0.05mm, as rolled, 99.99+%
Titanium, rod, 500mm, diameter 16mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 20mm, as drawn, 99.99+%
Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.95%
Titanium, microfoil, disks, 25mm, thinness 0.1μm, specific density 42.8μg/cm2, permanent mylar 3.5μm support, 99.6+%
Cadmium, foil, 6mm disks, thickness 0.01mm, 99.7+%
Titanium, tube, 200mm, outside diameter 1.05mm, inside diameter 0.75mm, wall thickness 0.15mm, annealed, 99.6+%
Cadmium, foil, 8mm disks, thickness 0.015mm, 99.7+%
Cadmium, foil, 50mm disks, thickness 0.125mm, as rolled, 99.99+%
Titanium, rod, 50mm, diameter 9.5mm, annealed, 99.99+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.01mm, 99.7+%
Cadmium, foil, light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 99.95%
Cadmium, tube, 100mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%