Merck
  • Attenuated aortic vasodilation and sympathetic prejunctional facilitation in epinephrine-deficient mice: selective impairment of β2-adrenoceptor responses.
Attenuated aortic vasodilation and sympathetic prejunctional facilitation in epinephrine-deficient mice: selective impairment of β2-adrenoceptor responses.
The Journal of pharmacology and experimental therapeutics (2014-08-28)
Mónica Moreira-Rodrigues, Ana L Graça, Marlene Ferreira, Joana Afonso, Paula Serrão, Manuela Morato, Fátima Ferreirinha, Paulo Correia-de-Sá, Steven N Ebert, Daniel Moura
ABSTRACT
It has been suggested that there is a link between epinephrine synthesis and the development of β2-adrenoceptor-mediated effects, but it remains to be determined whether this development is triggered by epinephrine. The aim of this study was to characterize β-adrenoceptor-mediated relaxation and facilitation of norepinephrine release in the aorta of phenylethanolamine-N-methyltransferase-knockout (Pnmt-KO) mice. Catecholamines were quantified by reverse-phase high-performance liquid chromatography-electrochemical detection. Aortic rings were mounted in a myograph to determine concentration-response curves to selective β1- or β2-adrenoceptor agonists in the absence or presence of selective β1- or β2-adrenoceptor antagonists. Aortic rings were also preincubated with [(3)H]norepinephrine to measure tritium overflow elicited by electrical stimulation in the presence of increasing concentrations of nonselective β- or selective β2-adrenoceptor agonists. β2-Adrenoceptor protein density was evaluated by Western blotting and β2-adrenoceptor localization by immunohistochemistry. Epinephrine is absent in Pnmt-KO mice. The potency and the maximal effect of the β2-adrenoceptor agonist terbutaline were lower in Pnmt-KO than in wild-type (WT) mice. The selective β2-adrenoceptor antagonist ICI 118,551 [(±)-erythro-(S*,S*)-1-[2,3-(dihydro-7-methyl-1H-inden-4-yl)oxy]-3-[(1-methylethyl)amino]-2-butanol hydrochloride] antagonized the relaxation caused by terbutaline in WT but not in Pnmt-KO mice. Isoproterenol and terbutaline induced concentration-dependent increases in tritium overflow in WT mice only. β2-Adrenoceptor protein density was decreased in membrane aorta homogenates of Pnmt-KO mice, and this finding was supported by immunofluorescence confocal microscopy. In conclusion, epinephrine is crucial for β2-adrenoceptor-mediated vasodilation and facilitation of norepinephrine release. In the absence of epinephrine, β2-adrenoceptor protein density was decreased in aorta cell membranes, thus potentially hindering its functional activity.
Sigma-Aldrich
L-Norepinephrine hydrochloride, ≥98.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
DL-Norepinephrine hydrochloride, crystalline, ≥97% (TLC)
Sigma-Aldrich
Phentolamine hydrochloride, ≥98% (TLC), powder
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline
Sigma-Aldrich
(±)-Norepinephrine (+)-bitartrate salt
Sigma-Aldrich
Terbutaline hemisulfate salt
Adrenaline, European Pharmacopoeia (EP) Reference Standard
Adrenaline with impurity F, European Pharmacopoeia (EP) Reference Standard
Isoprenaline sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Epinephrine
Sigma-Aldrich
Dobutamine hydrochloride, ≥98%
Sigma-Aldrich
Isoprenaline hydrochloride
Dobutamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Isoprenaline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Terbutaline sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DAPI, for nucleic acid staining