Merck
  • The limitations of diazepam as a treatment for nerve agent-induced seizures and neuropathology in rats: comparison with UBP302.
The limitations of diazepam as a treatment for nerve agent-induced seizures and neuropathology in rats: comparison with UBP302.
The Journal of pharmacology and experimental therapeutics (2014-08-27)
James P Apland, Vassiliki Aroniadou-Anderjaska, Taiza H Figueiredo, Franco Rossetti, Steven L Miller, Maria F M Braga
ABSTRACT
Exposure to nerve agents induces prolonged status epilepticus (SE), causing brain damage or death. Diazepam (DZP) is the current US Food and Drug Administration-approved drug for the cessation of nerve agent-induced SE. Here, we compared the efficacy of DZP with that of UBP302 [(S)-3-(2-carboxybenzyl)willardiine; an antagonist of the kainate receptors that contain the GluK1 subunit] against seizures, neuropathology, and behavioral deficits induced by soman in rats. DZP, administered 1 hour or 2 hours postexposure, terminated the SE, but seizures returned; thus, the total duration of SE within 24 hours after soman exposure was similar to (DZP at 1 hour) or longer than (DZP at 2 hours) that in the soman-exposed rats that did not receive the anticonvulsant. Compared with DZP, UBP302 stopped SE with a slower time course, but dramatically reduced the total duration of SE within 24 hours. Neuropathology and behavior were assessed in the groups that received anticonvulsant treatment 1 hour after exposure. UBP302, but not DZP, reduced neuronal degeneration in a number of brain regions, as well as neuronal loss in the basolateral amygdala and the CA1 hippocampal area, and prevented interneuronal loss in the basolateral amygdala. Anxiety-like behavior was assessed in the open field and by the acoustic startle response 30 days after soman exposure. The results showed that anxiety-like behavior was increased in the DZP-treated group and in the group that did not receive anticonvulsant treatment, but not in the UBP302-treated group. The results argue against the use of DZP for the treatment of nerve agent-induced seizures and brain damage and suggest that targeting GluK1-containing receptors is a more effective approach.
Sigma-Aldrich
L-Alanine, ≥99%
Pricing and availability is not currently available.
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Pricing and availability is not currently available.
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Thymine, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Thymine, suitable for cell culture, BioReagent
Pricing and availability is not currently available.
Supelco
Thymine(Zidovudine Related Compound C), Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Alanine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C
Pricing and availability is not currently available.
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
o-Xylene, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Pricing and availability is not currently available.
Supelco
o-Xylene, suitable for HPLC, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Pricing and availability is not currently available.
USP
Atropine sulfate, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Atropine sulfate salt monohydrate, ≥97% (TLC), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Pricing and availability is not currently available.
Supelco
Atropine Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.